Biphenyl-2,2'-dicarboxylic acid, Sigma-Aldrich®
Catalog # 8.02256.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:225 - 228 °C
- Molar Mass:242.23 g/mol
- Bulk Density:470 kg/m3
- pH:4 - 5 (H2O, 20 °C)
- Ignition Point:390 °C
- Synonyms:Diphenic acid, Diphenyl-2,2-dicarboxylic acid
- Hill Formula:C14H10O4
- Chemical Formula:2-(HOOC)C6H4C6H4-2-(COOH)
Specification Test Results
About this item
Synonyms: [1,1'-Biphenyl]-2,2'-dicarboxylic acid, Diphenic acid, 2,2'-Biphenyldicarboxylic acid
:
:242.23 g/mol
:M-Clarity™ quality level = MQ200
EINECS:207-576-4
CAS Number:482-05-3
Formula:C₁₄H₁₀O₄
MDL Number:MFCD00002464