
(S)-(-)-1-(1-Naphthyl)ethylamine 99+%
Catalog # ACRO217810010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.6215 to 1.6245 (20°C, 589 nm)
- GC:≥99.0 %
- Specific optical rotation:-51° to -59° (20°C, 589 nm) (c=2, ethanol)
Specification Test Results
About this item
:
:171.24 g/mol
Boiling Pt:153 °C (11 mmHg)
Density:1,06 g/cm³
Flash Pt:>112 °C
:GHS07
Storage Temperature:Ambient
CAS Number:10420-89-0
Formula:C₁₂H₁₃N
MDL Number:MFCD00064179