

(S)-(-)-1-(1-Naphthyl)ethylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10420-89-0
:
Formula:C₁₂H₁₃N
MDL Number:MFCD00064179
:171.24 g/mol
Boiling Pt:152…153 °C (11 mmHg)
Density:1,060
Flash Pt:>110 °C (230 °F)
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together