

(±)-1-(1-Naphthyl)ethylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:42882-31-5
:
Formula:C₁₂H₁₃N
MDL Number:MFCD00004014
:171.24 g/mol
Boiling Pt:155…156 °C (15 mmHg)
Density:1,063
Flash Pt:>110 °C (230 °F)
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together