
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:174899-66-2
:
Formula:C₉H₁₅F₃N₂O₃S
MDL Number:MFCD03427620
:288.29 g/mol
Melting Pt:16 °C
Density:1,295
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together