
Tryptamine hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:343-94-2
:
Formula:C₁₀H₁₃ClN₂
MDL Number:MFCD00012682
Merck Index:14,9796
Melting Pt:252…255 °C
:196.68 g/mol
EINECS:206-446-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together