Order Entry
Northern Ireland
ContactUsLinkComponent
Dicyclopropyl ketone, Sigma-Aldrich®
Flammable
Dicyclopropyl ketone, Sigma-Aldrich®
Catalog # 8.40157.0025
Supplier:  Merck
CAS Number:  
Flammable
Dicyclopropyl ketone, Sigma-Aldrich®
Catalog # 8.40157.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

Specification Test Results

  • Hill Formula:
    C₇H₁₀O
  • Molar Mass:
    110.15 g/mol
  • Boiling point:
    160 - 162 °C (1013 hPa)
  • Density:
    0.968 g/cm³
  • Flash point:
    39 °C
  • Storage class:
    3 Flammable liquids
  • WGK:
    WGK 3 highly water endangering
  • R Phrase:
    R 10

Specification Test Results

About this item

 :
 :110.16 g/mol
ADR:3,III
 :M-Clarity™ quality level = MQ200
UN:1993
 :GHS02
Storage Temperature:Ambient
CAS Number:1121-37-5
Formula:C₇H₁₀O
MDL Number:MFCD00001276