Order Entry
Northern Ireland
ContactUsLinkComponent
O-Dianisidine dihydrochloride, Supelco®
O-Dianisidine dihydrochloride, Supelco®
Catalog # D2679-1VL
Supplier:  Merck
CAS Number:  
O-Dianisidine dihydrochloride, Supelco®
Catalog # D2679-1VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :317.21
 :M-Clarity™ quality level = MQ100
EINECS:243-737-5
CAS Number:20325-40-0
Formula:C₁₄H₁₈Cl₂N₂O₂
MDL Number:MFCD00012488