Erythrosin B, red powder C.I. 45430:2
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:15905-32-5
:
Formula:C₂₀H₈I₄O₅
MDL Number:MFCD00005044
:835.89 g/mol
EINECS:240-474-8
:GHS07
Frequently Bought Together
Cryoboxes, Nalgene®
Supplier: Thermo Fisher Scientific
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Cell culture flasks, Falcon®
Supplier: Corning
Multiwell cell culture plates, Falcon®
Supplier: Corning
Cell culture plates, Nunc™ Multidish
Supplier: Thermo Fisher Scientific
Cryogenic tubes, CryoTubes™, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
VWR®, Cell Lifters
Supplier: VWR Collection
Microplates, 96-well
Supplier: Greiner Bio-One
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Colour coders for cryovials, CryoColorCode, Nunc™
Supplier: Thermo Fisher Scientific
Ethyl acetate, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Screw caps for laboratory/media bottles
Supplier: SciLabware