L-Threonine, Millipore®
Supplier: Merck
Certificates
About this item
CAS Number:72-19-5
:
Formula:C₄H₉NO₃
MDL Number:MFCD00064270
:119.12 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-774-1
Frequently Bought Together