Order Entry
Northern Ireland
ContactUsLinkComponent
 
2-(4-Toluoyl)benzoic acid, Sigma-Aldrich®
  8.42258.0025
 :  Merck
 :  
2-(4-Toluoyl)benzoic acid, Sigma-Aldrich®
  8.42258.0025
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Melting Point:
    133 - 137 °C
  • Molar Mass:
    240.26 g/mol

 

 

 :
 :240.26 g/mol
 :M-Clarity™ quality level = MQ200
 :201-614-3
 :GHS07
 :Ambient
 :85-55-2
 :C₁₅H₁₂O₃
 :MFCD00020287