
Hydroquinone monomethyl ether
Catalog # PROOMM1131.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:124.14 g/mol
Boiling Pt:243 °C (1013 hPa)
Melting Pt:55…57 °C
Density:1,55 g/cm³ (20 °C)
Flash Pt:128 °C
Storage Temperature:Ambient
EINECS:205-769-8
:GHS07
CAS Number:150-76-5
Formula:C₇H₈O₂
MDL Number:MFCD00002332