4-Nitroanisole 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-17-4
:
Formula:C₇H₇NO₃
MDL Number:MFCD00007327
Merck Index:12,06682
Boiling Pt:260 °C (1013 hPa)
Melting Pt:51…53 °C
Density:1,23 g/cm³
Flash Pt:130 °C
UN:3458
ADR:6.1,III
:153.14 g/mol
EINECS:202-825-3
:GHS08
Specification Test Results
- Appearance:Green to brown Crystals
- Infrared spectrum:Conforms
- Melting point:51 to 53 °C
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together