Order Entry
Northern Ireland
ContactUsLinkComponent
1,4-Dinitrobenzene for synthesis, Sigma-Aldrich®
1,4-Dinitrobenzene for synthesis, Sigma-Aldrich®
Catalog # 8.03120.0010
Supplier:  Merck
CAS Number:  
1,4-Dinitrobenzene for synthesis, Sigma-Aldrich®
Catalog # 8.03120.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    0.069 g/l (25 °C)
  • Melting Point:
    174 °C
  • Molar Mass:
    168.1 g/mol
  • Boiling Point:
    299 °C (1013 hPa)
  • Vapor Pressure:
    150 °C

Specification Test Results

About this item

 :
 :168.11 g/mol
Merck Index:13,03301
Boiling Pt:299 °C (1013 hPa)
Melting Pt:172 °C
Density:1,625 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-833-7
UN:3443
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS08
 :GHS09
CAS Number:100-25-4
Formula:C₆H₄N₂O₄
MDL Number:MFCD00007314