N-Methylaniline ≥98%
About this item
Purity: 98% CAS No.: 100-61-8 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 86°(186°F) Refractive Index: 1.5710 Melting Point: -57° Boiling Point: 194-197° Beilstein Reference: 741982 Merck Reference: 14,6019 UN No.: 2294 MDL No.: MFCD00008283
- CAS Number:100-61-8
- Formula:C₇H₉N
- MDL Number:MFCD00008283
- Molecular Weight:107.16 g/mol
- Merck Index:14,6019
- Boiling Pt:194…197 °C
- Melting Pt:–57 °C
- Density:0,989
- Flash Pt:86 °C (186 °F)
- Storage Temperature:Ambient
- EINECS:202-870-9
- UN:2294
- ADR:6.1,III
4 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 100-61-8 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 86°(186°F) Refractive Index: 1.5710 Melting Point: -57° Boiling Point: 194-197° Beilstein Reference: 741982 Merck Reference: 14,6019 UN No.: 2294 MDL No.: MFCD00008283
- CAS Number:100-61-8
- Formula:C₇H₉N
- MDL Number:MFCD00008283
- Molecular Weight:107.16 g/mol
- Merck Index:14,6019
- Boiling Pt:194…197 °C
- Melting Pt:–57 °C
- Density:0,989
- Flash Pt:86 °C (186 °F)
- Storage Temperature:Ambient
- EINECS:202-870-9
- UN:2294
- ADR:6.1,III
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



