Order Entry
Northern Ireland
ContactUsLinkComponent
Aniline, EMSURE® for analysis, Supelco®
Toxic
Corrosive
Health
Environment
Aniline, EMSURE® for analysis, Supelco®
Catalog # 1.01261.1000
Supplier:  Merck
CAS Number:  
undefined
Toxic
Corrosive
Health
Environment
Aniline, EMSURE® for analysis, Supelco®
Catalog # 1.01261.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    36 g/l (20 °C)
  • Melting Point:
    -6.2 °C
  • Molar Mass:
    93.13 g/mol
  • Boiling Point:
    184 °C (1013 hPa)
  • Vapor Pressure:
    0.5 hPa (20 °C)
  • Flash Point:
    76 °C
  • Refractive Index:
    1.5863 (20 °C, 589 nm)
  • Explosion Limit:
    1.2 - 11 %(V)
  • Density:
    1.02 g/cm3 (20 °C)
  • pH:
    8.8 (36 g/l, H2O, 20 °C)
  • Ignition Point:
    540 °C

Specification Test Results

About this item

 :
 :93.13 g/mol
Merck Index:13,00661
Boiling Pt:184 °C (1013 hPa)
Melting Pt:–6,2 °C
Density:1,019 g/cm³ (20 °C)
Flash Pt:76 °C
Storage Temperature:Ambient
EINECS:200-539-3
UN:1547
ADR:6.1,II
 :M-Clarity™ quality level = MQ300
 :GHS06
 :GHS05
 :GHS08
 :GHS09
CAS Number:62-53-3
Formula:C₆H₅NH₂
MDL Number:MFCD00007629