Water samplers, AquaSampler
Supplier: BURKLE
Certificates
About this item
Samplers available in three sizes.
- Ultra pure materials guaranteeing high purity
- Easy to clean with rounded edges and threads
- FEP/PTFE ultra pure and anti-adhesive surface
Designed for ground water samples from wells, pits, and other sources.
Frequently Bought Together