Order Entry
Northern Ireland
ContactUsLinkComponent
Methyl orange
Methyl orange
Catalog # MOLE42196193-500G
Supplier:  Molekula
CAS Number:  
Methyl orange
Catalog # MOLE42196193-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :327.34 g/mol
Merck Index:13,06127
Boiling Pt:∼90 °C (1013 hPa)
Melting Pt:> 300 °C
Density:∼1 g/cm³ (20 °C)
Flash Pt:38 °C
Storage Temperature:Ambient
EINECS:208-925-3
UN:3143
ADR:6.1,III
 :GHS06
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502