
1-Iodo-4-nitrobenzene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:636-98-6
:
Formula:C₆H₄INO₂
MDL Number:MFCD00007299
:249.01 g/mol
Boiling Pt:289 °C (772 mmHg)
Melting Pt:171…173 °C
Density:2,273 g/cm³ (20 °C)
Flash Pt:100 °C
EINECS:211-272-7
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to orange to brown Powder
- Infrared spectrum:Conforms
- Melting point:172 to 177 °C
- GC:≥98.5 %
- UV:at 270 nm A: ≤0.05
- pH:8 to 9
- Titration with HClO4:99.5 to 100.5 %
- Chloride (Cl):≤0.005 %
- Iron (Fe):≤0.0005 %
- Lead (Pb):≤0.0005 %
Specification Test Results
Frequently Bought Together