Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Iodoaniline
4-Iodoaniline
4-Iodoaniline
4-Iodoaniline
MOLE45378638-100G
:
Molekula
:
4-Iodoaniline
MOLE45378638-100G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
219.02 g/mol
:
13,05049
:
262 °C (1013 hPa)
:
61…64 °C
:
Ambient
:
208-743-4
:
GHS07
:
540-37-4
:
C₆H₆IN
:
MFCD00007848