Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Iodoaniline
3-Iodoaniline
3-Iodoaniline
MOLEM47329389
:
Molekula
:
3-Iodoaniline
MOLEM47329389
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
219.02 g/mol
:
271 °C (1013 hPa)
:
24…26 °C
:
1,821 g/cm³ (20 °C)
:
Refrigerator
:
210-922-7
:
GHS07
:
626-01-7
:
C₆H₆IN
:
MFCD00007781