

2-Iodoaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-43-0
:
Formula:C₆H₆IN
MDL Number:MFCD00007680
:219.02 g/mol
Boiling Pt:263 °C (1013 hPa)
Melting Pt:53…57 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:210-426-0
:GHS06
:GHS07
Related Information
Frequently Bought Together