
2-Iodoaniline 98%
:615-43-0
:
:C₆H₆IN
:MFCD00007680
:219.02 g/mol
:263 °C (1013 hPa)
:53…58 °C
:>110 °C
:Refrigerator
:210-426-0
:GHS07
- Appearance:White to yellow to beige to brown Powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:53 to 58 °C
- GC:≥97.5 %
Frequently Bought Together