Order Entry
Northern Ireland
ContactUsLinkComponent
4-Iodobenzoic acid 98%
Irritant
4-Iodobenzoic acid 98%
Certificates
undefined
Irritant
4-Iodobenzoic acid 98%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    270 to 274 °C
  • HPLC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together