

2,3,5,6-Tetrafluorophenol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:769-39-1
:
Formula:C₆H₂F₄O
MDL Number:MFCD00002157
:166.07 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:33…38 °C
Flash Pt:79 °C
EINECS:212-209-6
UN:2923
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:White Low melting crystalline solid
- Infrared spectrum:Conforms
- Melting point:33 to 38 °C
- GC:≥97.5 %
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together