Order Entry
Northern Ireland
Orders LinkContactUsLinkComponent
2-Fluorophenol 98%

2-Fluorophenol 98%

Irritant
Flammable
2-Fluorophenol 98%
Certificates
2-Fluorophenol 98%

3 Options Available Below

Product Details & Documents
Synonyms:  o-Fluorophenol
  • CAS Number:367-12-4
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002155
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:152 °C (760 mmHg)
  • Melting Pt:16,1 °C
  • Density:1,256 g/cm³
  • Flash Pt:46 °C
  • Storage Temperature:Ambient
  • EINECS:206-681-2
  • UN:1993
  • ADR:3,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow-brown Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5134 to 1.5154 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • UV:
    at 230 nm A: ≤0.2
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤2 ppm
  • Acidity:
    ≤0.0003 meq/g
  • Filtered to 0.2 micrometer:
    Filtered
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - £39.80
    £39.80
    1 * 10 g
       £39.80

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - £222.00
    £222.00
    1 * 100 g
       £222.00

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - £1,050.00
    £1,050.00
    1 * 500 g
       £1,050.00