Order Entry
Northern Ireland
ContactUsLinkComponent
1-Fluoro-2-nitrobenzene 99%
Toxic
Health
1-Fluoro-2-nitrobenzene 99%
Certificates
undefined
Toxic
Health
1-Fluoro-2-nitrobenzene 99%

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5307 to 1.5327 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together