1,2-Difluoro-4-nitrobenzene 99%
Catalog # ACRO241930250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5080 to 1.5100 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
About this item
:
:159.09 g/mol
Boiling Pt:80…81 °C (14 mmHg)
Density:1,441 g/cm³
Flash Pt:80 °C
Storage Temperature:Ambient
EINECS:206-718-2
UN:2810
ADR:6.1,III
:GHS07
CAS Number:369-34-6
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00007198