



2,3,4-Trifluoroaniline
Catalog # 8.41150.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 ml
- Seq:2070
Specifications
About this item
:
:147.1 g/mol
Boiling Pt:174 °C (1013 hPa)
Melting Pt:14…15 °C
Density:1,39 g/cm³ (20 °C)
EINECS:407-170-9
UN:3082
ADR:9,III
:GHS05
:GHS08
:GHS07
:GHS09
CAS Number:3862-73-5
Formula:C₆H₄F₃N
MDL Number:MFCD00011737