1,2-Difluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:367-11-3
:
Formula:C₆H₄F₂
MDL Number:MFCD00000284
:114.09 g/mol
Boiling Pt:92…94 °C
Melting Pt:–34 °C
Density:1,171
Flash Pt:1 °C (33 °F)
Storage Temperature:Ambient
EINECS:206-680-7
UN:1993
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together