
2,5-Dichlorothiophenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5858-18-4
:
Formula:C₆H₄Cl₂S
MDL Number:MFCD00004831
:179.07 g/mol
Boiling Pt:112…116 °C (50 mmHg)
Melting Pt:22…25 °C
Density:1,415 g/cm³ (30 °C)
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:227-486-9
UN:2810
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together