ContactUsLinkComponent
Safety spectacles, Cobra HD
  111-0620
undefined
Safety spectacles, Cobra HD
  111-0620
  • Type:
    Anti-UV (2C-1,2), anti-fog, anti-scratch
  • Colour:
    Black
  • Lenses:
    Clear PC (HD lens)
  • Pk:
    1
Cobra spectacles are multipurpose eyewear with a panoramic visual field. The side arms can be exchanged by a head band.
  • Non-slip bridge and temples
  • Clear, coated PC lens with anti-fog and anti-scratch properties for 180° view
  • HD lens with the highest level of optical clarity and aquaphobic properties
Weight: 25 g
Weight: 25 g
 : EN 166, EN 170
 : The Cobra spectacles are also available with a foam inner layer and head band (rather than arms), which allows them to be rated as a goggle for protection against liquid droplets.