


2,6-Dichloroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:608-31-1
:
Formula:C₆H₅Cl₂N
MDL Number:MFCD00007675
:162.02 g/mol
Boiling Pt:97 °C (5 mmHg)
Melting Pt:37…40 °C
Density:1,28
Flash Pt:118 °C (244 °F)
Storage Temperature:Ambient
EINECS:210-160-5
UN:3442
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Related Information
Frequently Bought Together