


3-Chloro-4-fluoroaniline ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:367-21-5
:
Formula:C₆H₅ClFN
MDL Number:MFCD00007767
:145.56 g/mol
Boiling Pt:227…230 °C
Melting Pt:44…47 °C
Density:1,420
Flash Pt:149 °C (300 °F)
Storage Temperature:Ambient
EINECS:206-682-8
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together