Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2-fluorobenzoic acid
4-Chloro-2-fluorobenzoic acid
4-Chloro-2-fluorobenzoic acid
MOLEM75667669
:
Molekula
:
4-Chloro-2-fluorobenzoic acid
MOLEM75667669
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
174.56 g/mol
:
207-162-3
:
GHS07
:
Ambient
:
446-30-0
:
C₇H₄ClFO₂
:
MFCD00042468