
2,4,6-Trichlorobenzoic acid ≥94%
Catalog # L14191.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:50-43-1
:
Formula:C₇H₃Cl₃O₂
:225.46 g/mol
Melting Pt:162…164 °C
:GHS07
Storage Temperature:Ambient
MDL Number:MFCD00060699