Order Entry
Northern Ireland
ContactUsLinkComponent
2-Chlorobenzonitrile 99%
Irritant
2-Chlorobenzonitrile 99%
Certificates
undefined
Irritant
2-Chlorobenzonitrile 99%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder or crystals and/or chunks or solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    43 to 46 °C
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together