

1,2-Dichlorobenzene 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-50-1
:
Formula:C₆H₄Cl₂
MDL Number:MFCD00000535
:147 g/mol
Merck Index:13,03081
Boiling Pt:179…180 °C (760 mmHg)
Melting Pt:–15 °C
Density:1,3 g/cm³
Flash Pt:67 °C
Storage Temperature:Ambient
EINECS:202-425-9
UN:1591
ADR:6.1,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5500 to 1.5520 (20°C, 589 nm)
- GC:≥98.5 %
- Suitability:for use as a laser dye
- Molar extinction coefficient:≥105000 (lambda max)
Specification Test Results
Frequently Bought Together