Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
1,2,4,5-Tetrachlorobenzene 98%
 
undefined
 
1,2,4,5-Tetrachlorobenzene 98%

 

  • Appearance:
    Light pink to grey Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    139 to 142 °C
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together