


Salicylic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:69-72-7
:
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439
:138.12 g/mol
Merck Index:13,08411
Boiling Pt:336 °C (1013 hPa)
Melting Pt:157…159 °C
Density:1,443 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:157 °C
Storage Temperature:Ambient
EINECS:200-712-3
:GHS05
:GHS08
:GHS07
Specification Test Results
- Solubility:2 g/l (20 °C)
- Melting Point:157 - 159 °C
- Molar Mass:138.12 g/mol
- Bulk Density:400 - 500 kg/m3
- Boiling Point:211 °C (1013 hPa)
- Vapor Pressure:27 hPa (211 °C)
- Flash Point:157 °C
- Density:1.443 g/cm3 (20 °C)
- pH:3 (H2O, 20 °C) (saturated solution)
- Ignition Point:500 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.18731.0100_TDS_EN ProductPage_818731
Product specification
msds_818731
SDS
Frequently Bought Together