Syringic acid for synthesis, Sigma-Aldrich®
Catalog # 8.22161.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:204 - 207 °C
- Molar Mass:198.18 g/mol
- Synonyms:3,5-Dimethoxy-4-hydroxybenzoic acid, Syringic acid
- Hill Formula:C9H10O5
- Chemical Formula:3,5-(CH3O)2-4-(OH)C6H2COOH
Specification Test Results
About this item
:
:198.18 g/mol
Boiling Pt:351 °C (1013 hPa)
Melting Pt:204…207 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:208-486-8
CAS Number:530-57-4
Formula:C₉H₁₀O₅
MDL Number:MFCD00002552