
4-Hydroxy-3,5-dinitrobenzoic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1019-52-9
:
Formula:C₇H₄N₂O₇
MDL Number:MFCD00017000
:228.12 g/mol
Melting Pt:248…252 °C
EINECS:213-814-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together