
4-(Methylamino)benzoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10541-83-0
:
Formula:C₈H₉NO₂
MDL Number:MFCD00002535
:151.16 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:158…160 °C
Storage Temperature:Ambient
EINECS:234-121-7
:GHS07
Related Information
Frequently Bought Together