
Sodium benzoate, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:532-32-1
:
Formula:C₆H₅COONa
MDL Number:MFCD00012463
:144.11 g/mol
Merck Index:13,08654
Melting Pt:410…430 °C
Density:1,44 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:208-534-8
REACH:01-2119460683-35
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (dried substance):99.0 - 100.5 %
- Alkalinity:passes test
- Heavy Metals (as Pb):max. 0.001 %
- Identification:passes test
- Organic Volatile Impurities:passes test
Specification Test Results
Frequently Bought Together