Order Entry
Northern Ireland
ContactUsLinkComponent
Isopentyl benzoate for synthesis, Sigma-Aldrich®
Isopentyl benzoate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Isopentyl benzoate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    192.25 g/mol
  • Boiling Point:
    260 °C
  • Flash Point:
    89 °C
  • Refractive Index:
    1.4949 (20 °C, 589 nm)
  • Density:
    0.99 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together