Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dimethoxybenzaldehyde
2,3-Dimethoxybenzaldehyde
2,3-Dimethoxybenzaldehyde
MOLE25022748-100G
:
Molekula
:
2,3-Dimethoxybenzaldehyde
MOLE25022748-100G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
166.18 g/mol
:
267 °C (1013 hPa)
:
48…49 °C
:
Ambient
:
201-677-7
:
86-51-1
:
C₉H₁₀O₃
:
MFCD00003309