Order Entry
Northern Ireland
ContactUsLinkComponent
2-Hydroxy-m-anisaldehyde
2-Hydroxy-m-anisaldehyde
Catalog # MOLE16781640-100G
Supplier:  Molekula
CAS Number:  
2-Hydroxy-m-anisaldehyde
Catalog # MOLE16781640-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.15 g/mol
Boiling Pt:265 °C (1013 hPa)
Melting Pt:40…42 °C
Density:1,2039 g/cm³ (45 °C)
Storage Temperature:Ambient
EINECS:205-715-3
CAS Number:148-53-8
Formula:C₈H₈O₃
MDL Number:MFCD00003322