Zip bags, double pocket without writing area, Corning® Gosselin™
Supplier: Corning
Certificates
About this item
Double pocket ZIP polyethylene bags.
- Non sterile
- Quick closure system for efficient transport of samples
Specifications
- Catalog No:
- GOSSSP2-01
- GOSSSP8-01
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Borre
- RECs, ISO 14001 - Borre