Distilling receivers, Anschütz-Thiele, straight, with cooling jacket
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
Made of DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Receivers with straight adapter, with cooling jacket, graduated, with socket and cone NS 29/32, vacuum connection: thread GL18, NS stopcock with solid glass or PTFE plug.
Frequently Bought Together