
2,4-Dibromo-6-nitroaniline 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:827-23-6
:
Formula:C₆H₄Br₂N₂O₂
MDL Number:MFCD00012107
:295.92 g/mol
Melting Pt:129…131 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together